Merck
  • Total synthesis and conformational analysis of the antifungal agent (-)-PF1163B.
Total synthesis and conformational analysis of the antifungal agent (-)-PF1163B.
Organic letters (2003-10-24)
Fodil Bouazza, Brigitte Renoux, Christian Bachmann, Jean-Pierre Gesson
ABSTRACT
[reaction: see text]. (-)-PF1163B, a new macrocyclic antifungal antibiotic isolated from Streptomyces sp., has been prepared in eight steps from (S)-citronellene. The key step is a ring-closing metathesis reaction of an ester and amide derivative obtained from a substituted N-methyl-l-tyrosine.
Sigma-Aldrich
Benzyl 2-bromoethyl ether, 97%
Pricing and availability is not currently available.