Merck
  • Dexmedetomidine Inhibits Maturation and Function of Human Cord Blood-Derived Dendritic Cells by Interfering with Synthesis and Secretion of IL-12 and IL-23.
Dexmedetomidine Inhibits Maturation and Function of Human Cord Blood-Derived Dendritic Cells by Interfering with Synthesis and Secretion of IL-12 and IL-23.
PloS one (2016-04-08)
Gong Chen, Yuan Le, Lei Zhou, Li Gong, Xiaoxiao Li, Yunli Li, Qin Liao, Kaiming Duan, Jianbin Tong, Wen Ouyang
ABSTRACT
To investigate the effects and underlying mechanism of dexmedetomidine on the cultured human dendritic cells (DCs). Human DCs and cytotoxic T lymphocytes (CTLs) were obtained from human cord blood mononuclear cells by density gradient centrifugation. Cultured DCs were divided into three groups: dexmedetomidine group, dexmedetomidine plus yohimbine (dexmedetomidine inhibitor) group and control group. DCs in the three groups were treated with dexmedetomidine, dexmedetomidine plus yohimbine and culture medium, respectively. After washing, the DCs were co-incubated with cultured CTLs. The maturation degree of DCs was evaluated by detecting (1) the ratios of HLA-DR-, CD86-, and CD80-positive cells (flow cytometry), and (2) expression of IL-12 and IL-23 (PCR and Elisa). The function of DCs was evaluated by detecting the proliferation (MTS assay) and cytotoxicity activity (the Elisa of IFN-γ) of CTLs. In addition, in order to explore the mechanisms of dexmedetomidine modulating DCs, α2-adrenergic receptor and its downstream signals in DCs were also detected. The ratios of HLA-DR-, CD86-, and CD80-positive cells to total cells were similar among the three groups (P>0.05). Compared to the control group, the protein levels of IL-12 and IL-23 in the culture medium and the mRNA levels of IL-12 p35, IL-12 p40 and IL-23 p19 in the DCs all decreased in dexmedetomidine group (P<0.05). In addition, the proliferation of CTLs and the secretion of IFN-γ also decreased in the dexmedetomidine group, compared with the control group (P<0.05). Moreover, these changes induced by dexmedetomidine in the dexmedetomidine group were reversed by α2-adrenergic receptor inhibitor yohimbine in the dexmedetomidine plus yohimbine group. It was also found the decrease of mRNA levels of IL-12 p35, IL-12 p40 and IL-23 p19 in the dexmedetomidine group could be reversed by ERK1/2 or AKT inhibitors. Dexmedetomidine could negatively modulate human immunity by inhibiting the maturation of DCs and then decreasing the proliferation and cytotoxicity activity of CTLs. The α2-adrenergic receptors and its downstream molecules ERK1/2 and AKT are closely involved in the modulation of dexmedetomidine on DCs.
Sigma-Aldrich
PD 98,059, solid
Sigma-Aldrich
LY-294,002 hydrochloride, solid, ≥98% (HPLC)
Sigma-Aldrich
SB 203580, solid, ≥98% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate–dextran, average mol wt 40,000
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution