Merck
  • An improved method for the palladium-catalyzed amination of aryl iodides.
An improved method for the palladium-catalyzed amination of aryl iodides.
The Journal of organic chemistry (2001-04-17)
M H Ali, S L Buchwald
ABSTRACT
Aryl iodides are coupled with amines to give the corresponding arylamines in high yield in the presence of palladium, a suitable ligand, and NaOt-Bu. Functionalized aryl iodides give good yields of the corresponding arylamines when Cs(2)CO(3) is substituted as the base.
Sigma-Aldrich
1-Iodo-3,5-dimethylbenzene, 99%