Merck
  • Beat Rate Variability in Murine Embryonic Stem Cell-Derived Cardiomyocytes: Effect of Antiarrhythmic Drugs.
Beat Rate Variability in Murine Embryonic Stem Cell-Derived Cardiomyocytes: Effect of Antiarrhythmic Drugs.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2016-02-06)
Julius Niehoff, Matthias Matzkies, Filomain Nguemo, Jürgen Hescheler, Michael Reppel
ABSTRACT
Heart rate variability (HRV) refers to the fluctuation of the time interval between consecutive heartbeats in humans. It has recently been discovered that cardiomyocytes derived from human embryonic and induced pluripotent stem cells show beat rate variability (BRV) that is similar to the HRV in humans. In the present study, clinical aspects of HRV were transferred to an in vitro model. The aims of the study were to explore the BRV in murine embryonic stem cell (mESC)-derived cardiomyocytes and to demonstrate the influence of antiarrhythmic drugs on BRV as has been shown in clinical trials previously. The Microelectrode Array (MEA) technique was used to perform short-term recordings of extracellular field potentials (FPs) of spontaneously beating cardiomyocytes derived from mESCs (D3 cell line, αPig-44). Offline analysis was focused on time domain and nonlinear methods. The Poincaré-Plot analysis of measurements without pharmacological intervention revealed that three different shapes of scatter plots occurred most frequently. Comparable shapes have been described in clinical studies before. The antiarrhythmic drugs Ivabradine, Verapamil and Sotalol augmented BRV, whereas Flecainide decreased BRV parameters at low concentrations (SDSD 79.0 ± 8.7% of control at 10(-9) M, p < 0.05) and increased variability measures at higher concentrations (SDNN 258.8 ± 42.7% of control at 10(-5) M, p < 0.05). Amiodarone and Metoprolol did not alter BRV significantly. Spontaneously beating cardiomyocytes derived from mESCs showed BRV that appears to be similar to the HRV known from humans. Antiarrhythmic drugs affected BRV parameters similar to clinical observations. Therefore, our study demonstrates that this in vitro model can contribute to a better understanding of electrophysiological properties of mESC-derived cardiomyocytes and might serve as a valuable tool for drug safety screening.
Sigma-Aldrich
Ivabradine hydrochloride, ≥98% (HPLC)
Pricing and availability is not currently available.