Merck
  • Enhanced lymph vessel density, remodeling, and inflammation are reflected by gene expression signatures in dermal lymphatic endothelial cells in type 2 diabetes.
Enhanced lymph vessel density, remodeling, and inflammation are reflected by gene expression signatures in dermal lymphatic endothelial cells in type 2 diabetes.
Diabetes (2013-02-21)
Monika Haemmerle, Thomas Keller, Gerda Egger, Helga Schachner, Carl Walter Steiner, Dejan Stokic, Christoph Neumayer, Markus K Brown, Dontscho Kerjaschki, Brigitte Hantusch
ABSTRACT
Type 2 diabetes is associated with microvascular damage that causes frequent infections in the skin and chronic ulcers as a result of impaired wound healing. To trace the pathological changes, we performed a comprehensive analysis of lymphatic vessels in the skin of type 2 diabetic versus nondiabetic patients. The dermis revealed enhanced lymphatic vessel density, and transcriptional profiling of ex vivo isolated lymphatic endothelial cells (LECs) identified 160 genes differentially expressed between type 2 diabetic and nondiabetic LECs. Bioinformatic analysis of deregulated genes uncovered sets functionally related to inflammation, lymphatic vessel remodeling, lymphangiogenesis, and lipid and small molecule transport. Furthermore, we traced CD68(+) macrophage accumulation and concomitant upregulation of tumor necrosis factor-α (TNF-α) levels in type 2 diabetic skin. TNF-α treatment of LECs and its specific blockade in vitro reproduced differential regulation of a gene set that led to enhanced LEC mobility and macrophage attachment, which was mediated by the LEC-derived chemokine CXCL10. This study identifies lymph vessel gene signatures directly correlated with type 2 diabetes skin manifestations. In addition, we provide evidence for paracrine cross-talk fostering macrophage recruitment to LECs as one pathophysiological process that might contribute to aberrant lymphangiogenesis and persistent inflammation in the skin.
Product Description
Roche
Dispase® I (neutral protease, grade I), lyophilized, ≥10 units/mg protein (20 U/vial (37 °C, casein as substrate, pH 7.5)), optimum pH 6.0-8.5
Pricing and availability is not currently available.
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Pricing and availability is not currently available.