Merck
  • Mimicking a Stenocara beetle's back for microcondensation using plasmachemical patterned superhydrophobic-superhydrophilic surfaces.
Mimicking a Stenocara beetle's back for microcondensation using plasmachemical patterned superhydrophobic-superhydrophilic surfaces.
Langmuir : the ACS journal of surfaces and colloids (2007-01-11)
R P Garrod, L G Harris, W C E Schofield, J McGettrick, L J Ward, D O H Teare, J P S Badyal
ABSTRACT
A simple two-step plasmachemical methodology is outlined for the fabrication of microcondensor surfaces. This comprises the creation of a superhydrophobic background followed by pulsed plasma deposition of a hydrophilic polymer array. Microcondensation efficiency has been explored in terms of the chemical nature of the hydrophilic pixels and their dimensions. These results are compared to the hydrophilic-hydrophobic pattern present on the Stenocara beetle's back, which is used by the insect to collect water in the desert. Potential applications include fog harvesting, microfluidics, and biomolecule immobilization.
Sigma-Aldrich
3-Vinylbenzaldehyde, 97%