Merck
  • Enantioselective Addition of Diethylzinc to Aldehydes Catalyzed by Chiral O,N,O-tridentate Phenol Ligands Derived From Camphor.
Enantioselective Addition of Diethylzinc to Aldehydes Catalyzed by Chiral O,N,O-tridentate Phenol Ligands Derived From Camphor.
Chirality (2015-10-22)
Dong-Sheng Lee, Shu-Ming Chang, Chun-Ying Ho, Ta-Jung Lu
ABSTRACT
Chiral O,N,O-tridentate phenol ligands bearing a camphor backbone were found to be effective chiral catalysts for the enantioselective addition of diethylzinc to aromatic aldehydes, resulting in high enantioselectivities (80-95% ee) at room temperature.
Sigma-Aldrich
2-Naphthaldehyde, 98%