Merck
  • In-vivo transfection of pcDNA3.1-IGFBP7 inhibits melanoma growth in mice through apoptosis induction and VEGF downexpression.
In-vivo transfection of pcDNA3.1-IGFBP7 inhibits melanoma growth in mice through apoptosis induction and VEGF downexpression.
Journal of experimental & clinical cancer research : CR (2010-02-18)
Rong-Yi Chen, Hong-Xiang Chen, Jia-Xi Lin, Wei-Bing She, Ping Jiang, Li Xu, Ya-Ting Tu
ABSTRACT
Genome-wide RNA interference screening study revealed that loss of expression of insulin-like growth factor binding protein 7 (IGFBP7) is a critical step in development of a malignant melanoma (MM), and this secreted protein plays a central role in apoptosis of MM. In this study we constructed pcDNA3.1-IGFBP7 to obtain high expression of IGBPF7 and to inhibit the growth of MM in C57BL/6J mice. pcDNA3.1-IGFBP7 was transfected into B16-F10 cell, the expression of IGFBP7 was detected by RT-PCR and western blot. The proliferations and apoptosis rates of transfected and control cells were measured by CCK8 and FCM, respectively. The tumorigenicity and tumor growth in both pcDNA3.1-IGFBP7 group and control groups were studied in C57BL/6J mice model. IGFBP7, caspase-3, and VEGF expressions in tumor tissue were measured by immunohistochemistry. Apoptosis of tumors were detected by TUNEL. We demonstrated this plasmid inhibited proliferation of B16-F10 melanoma cells efficiently in vivo, exploiting the high expression of IGFBP7. More importantly, in-vivo transfection of pcDNA3.1-IGFBP7 inhibited MM growth in C57BL/6J mice. The inhibition of MM growth was proved owing to apoptosis and reduced expression of VEGF induced by pcDNA3.1-IGFBP7. These results suggest a potential new clinical strategy for MM gene treatment.
Product Description
Roche
In Situ Cell Death Detection Kit, AP, sufficient for ≤50 tests, kit of 1 (3 components), suitable for detection