Merck
  • Role of GDF15 (growth and differentiation factor 15) in pulmonary oxygen toxicity.
Role of GDF15 (growth and differentiation factor 15) in pulmonary oxygen toxicity.
Toxicology in vitro : an international journal published in association with BIBRA (2015-05-26)
Kirti Kumar Tiwari, Bhagavatula Moorthy, Krithika Lingappan
ABSTRACT
GDF15 (growth and differentiation factor 15) is a secreted cytokine, a direct target of p53 and plays a role in cell proliferation and apoptosis. It is induced by oxidative stress and has anti-apoptotic effects. The role of GDF15 in hyperoxic lung injury is unknown. We tested the hypothesis that GDF15 will be induced in vitro, in a model of pulmonary oxygen toxicity, and will play a critical role in decreasing cell death and oxidative stress. BEAS-2B (human bronchial epithelial cells) and human pulmonary vascular endothelial cells (HPMEC) were exposed to hyperoxia, and expression of GDF15 and effect of GDF15 disruption on cell viability and oxidative stress was determined. Furthermore, we studied the effect of p53 knockdown on GDF15 expression. In vitro, both BEAS-2B and HPMEC cells showed a significant increase in GDF15 expression upon exposure to hyperoxia. After GDF15 knockdown, there was a significant decrease in cell viability and increase in oxidative stress compared to control cells transfected with siRNA with a scrambled sequence. Knockdown of p53 significantly decreased the induction of GDF15 by hyperoxia. In conclusion, we show that GDF15 has a pro-survival and anti-oxidant role in hyperoxia and that p53 plays a key role in its induction.
Sigma-Aldrich
MISSION® esiRNA, targeting human GDF15
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Gdf15
Sigma-Aldrich
MISSION® esiRNA, targeting human TP53