Merck
  • Negative regulation of the LKB1/AMPK pathway by ERK in human acute myeloid leukemia cells.
Negative regulation of the LKB1/AMPK pathway by ERK in human acute myeloid leukemia cells.
Experimental hematology (2015-04-08)
Ichiro Kawashima, Toru Mitsumori, Yumi Nozaki, Takeo Yamamoto, Yuki Shobu-Sueki, Kei Nakajima, Keita Kirito
ABSTRACT
Adenosine monophosphate-activated protein kinase (AMPK) is a sensor for cellular energy status. When the cellular energy level is decreased, AMPK is activated and functions to suppress energy-consuming processes, including protein synthesis. Recently, AMPK has received attention as an attractive molecular target for cancer therapy. Several studies have revealed that the activation of AMPK by chemical stimulators, such as metformin, induces apoptosis in a variety of hematologic malignant cells. From another perspective, these results suggest that the function of AMPK is impaired in hematologic tumor cells. However, the precise mechanisms by which this impairment occurs are not well understood. In melanoma cells, oncogenic BRAF constitutively activates the extracellular signal-regulated kinase (ERK) pathway and phosphorylates liver kinase B1, an upstream activator of 5' adenosine monophosphate-activated protein kinase (AMPK), resulting in the inactivation of liver kinase B1 and AMPK. In this study, we analyzed whether ERK is involved in the suppression of AMPK activity using established and primary human leukemia cells. We found an inverse correlation between the intensity of ERK activity and the degree of AMPK activation after stimulation with either glucose deprivation or metformin. We also found that the inhibition of ERK activity by U0126 restored AMPK activation after metformin treatment. Furthermore, a combined treatment with metformin and U0126 enhanced the antileukemic activity of metformin. Importantly, metformin induced ERK activation by suppressing the protein levels of dual specificity phosphatase 6, a negative regulator of ERK. This crosstalk between AMPK and ERK could diminish the antileukemic activity of metformin. Taken together, our present observations suggest a novel therapeutic strategy for improving the efficacy of metformin in treating leukemia.
Sigma-Aldrich
MISSION® esiRNA, targeting human PRKAA1
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
DL-Tyrosine, 99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Prkaa1