Merck
  • New t-butyl based aspartate protecting groups preventing aspartimide formation in Fmoc SPPS.
New t-butyl based aspartate protecting groups preventing aspartimide formation in Fmoc SPPS.
Journal of peptide science : an official publication of the European Peptide Society (2015-06-17)
Raymond Behrendt, Simon Huber, Roger Martí, Peter White
ABSTRACT
Obtaining homogenous aspartyl-containing peptides via Fmoc/tBu chemistry is often an insurmountable obstacle. A generic solution for this issue utilising an optimised side-chain protection strategy that minimises aspartimide formation would therefore be most desirable. To this end, we developed the following new derivatives: Fmoc-Asp(OEpe)-OH (Epe = 3-ethyl-3-pentyl), Fmoc-Asp(OPhp)-OH (Php = 4-n-propyl-4-heptyl) and Fmoc-Asp(OBno)-OH (Bno = 5-n-butyl-5-nonyl). We have compared their effectiveness against that of Fmoc-Asp(OtBu)-OH and Fmoc-Asp(OMpe)-OH in the well-established scorpion toxin II model peptide variants H-Val-Lys-Asp-Asn/Arg-Tyr-Ile-OH by treatments of the peptidyl resins with the Fmoc removal reagents containing piperidine and DBU at both room and elevated temperatures. The new derivatives proved to be extremely effective in minimising aspartimide by-products in each application.
Sigma-Aldrich
Fmoc-Asp(OtBu)-OH, ≥98.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Fmoc-Asp(OBno)-OH, Novabiochem®
Pricing and availability is not currently available.
Sigma-Aldrich
3-Ethyl-3-pentanol, 98%
Pricing and availability is not currently available.