Merck
  • Serological and Molecular Investigation of Swine Hepatitis E Virus in Pigs Raised in Southern Italy.
Serological and Molecular Investigation of Swine Hepatitis E Virus in Pigs Raised in Southern Italy.
Journal of food protection (2015-11-12)
Nicola Costanzo, Eleonora Sarno, Vincenzo Peretti, Lucia Ciambrone, Francesco Casalinuovo, Adriano Santoro
ABSTRACT
Hepatitis E virus (HEV) infection is a common acute hepatitis transmitted by the fecal-oral route. In developed countries, the virus has a zoonotic potential, and domestic pigs and wild boars are considered main reservoirs. To assess the prevalence of HEV-positive animals in the Calabria region (southern Italy) on a serological and molecular level, a total of 216 autochthonous healthy pigs (Apulo-Calabrese breed) were sampled. Both sera and feces were collected. Pigs were grouped based on age: 117 pigs <6 months and 99 pigs >6 months. By using a commercial enzyme-linked immunosorbent assay system, a total of 173 (80%) of the 216 pigs tested seropositive. In all sampled farms (n = 8), pigs with antibodies (immunoglobulin G) against HEV were detected at a level higher than 60%, with a significant difference among age groups (P < 0.0001). Moreover, 16 fattening pigs were found to be nested reverse transcription PCR positive and thus to shed viral genomes in their feces. These positive findings resulted in a prevalence of 48.4% on the farm level (16 of 35 pigs) and an overall prevalence of 7.4% at the animal level (16 of 216 pigs). Based on the present study, HEV seems to circulate among the autochthonous domestic pig population of southern Italy with a low sharing rate. Further studies exploring the origin of infection are needed to minimize the risk of human exposure and to reduce consequences for public health.
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Diethyl pyrocarbonate, Vetec, reagent grade, ≥97%