Merck
  • Bio-Inspired Chemical Fabrication of Stretchable Transparent Electrodes.
Bio-Inspired Chemical Fabrication of Stretchable Transparent Electrodes.
Small (Weinheim an der Bergstrasse, Germany) (2015-03-20)
You Yu, Yaokang Zhang, Kan Li, Casey Yan, Zijian Zheng
ABSTRACT
Stretchable and transparent electrodes are fabricated by chemical deposition of metal thin films on natural veins of leaves at ambient conditions. These vein-based transparent electrodes show excellent electro-optical property (0.9 Ω sq(-1) at 83% T) even at 50% tensile strains, ideal for flexible and stretchable optoelectronic devices.
Sigma-Aldrich
Ammonium tetrachloropalladate(II), 97%
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium tetrachloropalladate(II), ≥99.995% trace metals basis
Pricing and availability is not currently available.