Merck
  • COX/mPGES-1/PGE2 pathway depicts an inflammatory-dependent high-risk neuroblastoma subset.
COX/mPGES-1/PGE2 pathway depicts an inflammatory-dependent high-risk neuroblastoma subset.
Proceedings of the National Academy of Sciences of the United States of America (2015-06-17)
Karin Larsson, Anna Kock, Helena Idborg, Marie Arsenian Henriksson, Tommy Martinsson, John I Johnsen, Marina Korotkova, Per Kogner, Per-Johan Jakobsson
ABSTRACT
The majority of solid tumors are presented with an inflammatory microenvironment. Proinflammatory lipid mediators including prostaglandin E2 (PGE2) contribute to the establishment of inflammation and have been linked to tumor growth and aggressiveness. Here we show that high-risk neuroblastoma with deletion of chromosome 11q represents an inflammatory subset of neuroblastomas. Analysis of enzymes involved in the production of proinflammatory lipid mediators showed that 11q-deleted neuroblastoma tumors express high levels of microsomal prostaglandin E synthase-1 (mPGES-1) and elevated levels of PGE2. High mPGES-1 expression also corresponded to poor survival of neuroblastoma patients. Investigation of the tumor microenvironment showed high infiltration of tumor-promoting macrophages with high expression of the M2-polarization markers CD163 and CD206. mPGES-1-expressing cells in tumors from different subtypes of neuroblastoma showed differential expression of one or several cancer-associated fibroblast markers such as vimentin, fibroblast activation protein α, α smooth muscle actin, and PDGF receptor β. Importantly, inhibition of PGE2 production with diclofenac, a nonselective COX inhibitor, resulted in reduced tumor growth in an in vivo model of 11q-deleted neuroblastoma. Collectively, these results suggest that PGE2 is involved in the tumor microenvironment of specific neuroblastoma subgroups and indicate that therapeutic strategies using existing anti-inflammatory drugs in combination with current treatment should be considered for certain neuroblastomas.
Sigma-Aldrich
Anti-Disialoganglioside GD2 Antibody, clone 14G2a, clone 14.2Ga, Chemicon®, from mouse
Sigma-Aldrich
Monoclonal Anti-S100A4 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0240, purified immunoglobulin, buffered aqueous glycerol solution