- Cloning and characterization of the human IFT20 gene.
Cloning and characterization of the human IFT20 gene.
Intraflagellar transport (IFT), in which molecular motors and IFT particle proteins participate, is very important in assembling and maintaining many cilia/flagella, such as the motile cilia that drive the swimming of cells and embryos, the nodal cilia that generate left-right asymmetry in vertebrate embryos, and the sensory cilia that detect sensory stimuli in some animals. Here we report the molecular cloning and characterization of a novel human cDNA that is 1060 base pair in length, encoding a putative protein of 158 amino acid residues which shares high homology to Mus musculus intraflagellar transport protein mIFT20 (Accession number AAL99202, identity 82.3%, similarity 82.9%) at protein level. Bioinformatics analysis revealed that the gene is composed of 6 exons and is located on human chromosome 17p 11.1. RT-PCR showed that this gene is expressed in human brain, lung, kidney and pancreas, and lower expression were also detected in human placenta, liver, thymus, prostate and testis.