Merck
  • Simultaneous targeting of Requiem & Alg-2 in Chinese hamster ovary cells for improved recombinant protein production.
Simultaneous targeting of Requiem & Alg-2 in Chinese hamster ovary cells for improved recombinant protein production.
Molecular biotechnology (2010-06-24)
Yiping Lim, Athanasios Mantalaris, Miranda G S Yap, Danny C F Wong
ABSTRACT
Apoptosis is known to be the main cause of cell death in the bioreactor environment, leading to the loss of recombinant protein productivity. In a previous study, transcriptional profiling was used to identify and target four early apoptosis-signaling genes: FADD, FAIM, Alg-2, and Requiem. The resulting cell lines had increased viable cell numbers and extended culture viability, which translated to increased protein productivity. Combinatorial targeting of two genes simultaneously has previously been shown to be more effective than targeting one gene alone. In this study, we sought to determine if targeting Requiem and Alg-2 was more effective than targeting Requiem alone. We found that targeting Requiem and Alg-2 did not result in extended culture viability, but resulted in an increase in maximum viable cell numbers and cumulative IVCD under fed-batch conditions. This in turn led to an approximately 1.5-fold increase in recombinant protein productivity.