Merck
Design and preparation of 2-benzamido-pyrimidines as inhibitors of IKK.
Bioorganic & medicinal chemistry letters (2005-10-21)
Rudolf Waelchli, Birgit Bollbuck, Christian Bruns, Thomas Buhl, Jörg Eder, Roland Feifel, Rene Hersperger, Philipp Janser, Laszlo Revesz, Hans-Günter Zerwes, Achim Schlapbach
ABSTRACT
The design, synthesis, and the biological evaluation of 2-benzamido-pyrimidines as novel IKK inhibitors are described. By optimization of the lead compound 3, compounds 16 and 24 are identified as good inhibitors of IKK2 with IC(50) values of 40 and 25 nM, respectively. Compound 16 also demonstrated significant in vivo activity in an acute model of cytokine release.
Sigma-Aldrich
IKK Inhibitor VII, The IKK Inhibitor VII, also referenced under CAS 873225-46-8, controls the biological activity of IKK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Pricing and availability is not currently available.