Merck
  • Discovery and computer aided potency optimization of a novel class of small molecule CXCR4 antagonists.
Discovery and computer aided potency optimization of a novel class of small molecule CXCR4 antagonists.
PloS one (2013-11-10)
Victoria Vinader, Djevdet S Ahmet, Mohaned S Ahmed, Laurence H Patterson, Kamyar Afarinkia
ABSTRACT
Amongst the chemokine signalling axes involved in cancer, chemokine CXCL12 acting on chemokine receptor CXCR4 is particularly significant since it orchestrates migration of cancer cells in a tissue-specific metastatic process. High CXCR4 tumour expression is associated with poor prognosis of lung, brain, CNS, blood and breast cancers. We have identified a new class of small molecule CXCR4 antagonists based on the use of computational modelling studies in concert with experimental determination of in vitro activity against CXCL12-induced intracellular calcium mobilisation, proliferation and chemotaxis. Molecular modelling proved to be a useful tool in rationalising our observed potencies, as well as informing the direction of the synthetic efforts aimed at producing more potent compounds.
Product Description
Corning® Transwell® polycarbonate membrane cell culture inserts, 6.5 mm Transwell Inserts with 8.0 μm pore polycarbonate membrane coated w/ Cultrex BME, w/lid, sterile, 24/cs
Corning® Transwell® polycarbonate membrane cell culture inserts, 6.5 mm Transwell with 8.0 μm pore polycarbonate membrane insert, TC-treated, w/lid, sterile, 48/cs