Merck
Structural analysis of hydroxyapatite coatings on titanium.
Biomaterials (1986-03-01)
P Ducheyne, W Van Raemdonck, J C Heughebaert, M Heughebaert
ABSTRACT
Hydroxyapatite from two sources was electrophoretically deposited onto flat titanium plate material. Depending upon the deposition conditions various changes in the structure of the ceramic were identified. A well-adhering Ti-P compound was present at the interface. Hydroxyapatite oxygenated to various degrees and tetracalcium phosphate were reproducibly formed in the coating.
Sigma-Aldrich
Hydroxyapatite, puriss., meets analytical specification of Ph. Eur., BP, FCC, E341, ≥90% (calculated on glowed substance)
Pricing and availability is not currently available.
Sigma-Aldrich
Hydroxyapatite, purum p.a., ≥90% (as Ca3(PO4)2, KT)
Pricing and availability is not currently available.