Merck
  • A novel alteration in metaxin 1, F202L, is associated with N370S in Gaucher disease.
A novel alteration in metaxin 1, F202L, is associated with N370S in Gaucher disease.
Journal of human genetics (2004-03-17)
Mary E LaMarca, Mona Goldstein, Nahid Tayebi, Mauricio Arcos-Burgos, Brian M Martin, Ellen Sidransky
ABSTRACT
The gene for glucocerebrosidase ( GBA), the enzyme deficient in Gaucher disease, is located in a gene-rich region on 1q21. Metaxin 1( MTX1) is a convergently transcribed gene contiguous to the 3' end of the GBA pseudogene. A single nucleotide alteration in MTX1, 628T-->C, resulting in the amino acid change F202L, was identified in patients with Gaucher disease in association with the common N370S mutation in GBA. The polymorphism was also present on 4.6% of 152 control alleles, but could have functional consequences that have a modifying role in Gaucher disease.