Merck
  • Boc groups as protectors and directors for Ir-catalyzed C-H borylation of heterocycles.
Boc groups as protectors and directors for Ir-catalyzed C-H borylation of heterocycles.
The Journal of organic chemistry (2009-11-10)
Venkata A Kallepalli, Feng Shi, Sulagna Paul, Edith N Onyeozili, Robert E Maleczka, Milton R Smith
ABSTRACT
Ir-catalyzed C-H borylation is found to be compatible with Boc protecting groups. Thus, pyrroles, indoles, and azaindoles can be selectively functionalized at C-H positions beta to N. The Boc group can be removed on thermolysis or left intact during subsequent transformations.
Sigma-Aldrich
N-Boc-pyrrole, 98%
Pricing and availability is not currently available.