Merck
  • A planar Schiff base platinum(II) complex: crystal structure, cytotoxicity and interaction with DNA.
A planar Schiff base platinum(II) complex: crystal structure, cytotoxicity and interaction with DNA.
Chemical & pharmaceutical bulletin (2014-03-04)
Yan Peng, Hui Zhong, Zhen-Feng Chen, Yan-Cheng Liu, Guo-Hai Zhang, Qi-Pin Qin, Hong Liang
ABSTRACT
A new platinum(II) complex of salphen derivative, namely Schiff base ligand that derived from o-phenylenediamine and 5-chlorosalicylaldehyde was synthesized. The complex possessed a planar mononuclear structure. The in vitro cytotoxicities of the complex were evaluated by microculture tetrozolium (MTT) assay against seven human tumor cell lines with the IC50 values of ca. 11.61 µM. Cell cycle analysis indicated that the complex induced apoptosis and G1-phase arrest in A549 cells. The results of colony formation assay showed that the complex could suppress the proliferation and viability of A549 cells. The binding of the complex to potential target DNA were investigated by fluorescence spectroscopy, viscosity measurements, fluorescence polarization and agarose gel electrophoresis. The results suggest that the most probable binding mode of the complex is intercalation.
Sigma-Aldrich
5-Chlorosalicylaldehyde, 98%