Merck
  • Isolation of nitrophenols from diesel exhaust particles (DEP) as vasodilatation compounds.
Isolation of nitrophenols from diesel exhaust particles (DEP) as vasodilatation compounds.
Biological & pharmaceutical bulletin (2003-03-04)
Yoki Mori, Kazuyuki Kamata, Noriko Toda, Hideyuki Hayashi, Koh-ichi Seki, Shinji Taneda, Shin Yoshino, Akiyo Sakushima, Masakatsu Sakata, Akira K Suzuki
ABSTRACT
The compounds in diesel exhaust particles (DEP) that are responsible for vasodilatation were isolated and characterized for the first time. From benzene extract of DEP, 2-methyl-4-nitrophenol, 3-methyl-4-nitrophenl and 4-nitrophenol were isolated, and their vasodilatation activities were confirmed. 3-methyl-4-nitrophenol caused dilatation of rat thoracic artery, and the other two nitrophenols, also showed vasodilatation activities.
Sigma-Aldrich
2-Methyl-4-nitrophenol, 97%
Pricing and availability is not currently available.