Merck
  • Albocollagenase, a novel recombinant P-III snake venom metalloproteinase from green pit viper (Cryptelytrops albolabris), digests collagen and inhibits platelet aggregation.
Albocollagenase, a novel recombinant P-III snake venom metalloproteinase from green pit viper (Cryptelytrops albolabris), digests collagen and inhibits platelet aggregation.
Toxicon : official journal of the International Society on Toxinology (2011-02-22)
Anuwat Pinyachat, Ponlapat Rojnuckarin, Chuanchom Muanpasitporn, Pon Singhamatr, Surang Nuchprayoon
ABSTRACT
Molecular cloning and functional characterization of P-III snake venom metalloproteinases (SVMPs) will give us deeper insights in the pathogenesis of viper bites. This may lead to novel therapy for venom-induced local tissue damages, the complication refractory to current antivenom. The aim of this study was to elucidate the in vitro activities of a new SVMP from the green pit viper (GPV) using recombinant DNA technology. We report, here, a new cDNA clone from GPV (Cryptelytrops albolabris) venom glands encoding 614 amino acid residues P-III SVMP, termed albocollagenase. The conceptually translated protein comprised a signal peptide and prodomain, followed by a metalloproteinase domain containing a zinc-binding motifs, HEXGHXXGXXH-CIM and 9 cysteine residues. The disintegrin-like and cysteine-rich domains possessed 24 cysteines and a DCD (Asp-Cys-Asp) motif. The albocollagenase deduced amino acid sequence alignments showed approximately 70% identity with other P-III SVMPs. Notably, the prodomain was highly conserved, while the metalloproteinase, disintegrin-like and cysteine-rich domains contained several differences. Albocollagenase without the signal peptide and prodomain was expressed in Pichia pastoris with an N-terminal six-histidine tag. After affinity purification from the supernatant of methanol-induced media, SDS-PAGE and Western blot analysis in both reducing and non-reducing conditions showed a protein band of approximately 62 kDa. The recombinant albocollagenase could digest human type IV collagen from human placenta basement membrane within 1 min. After 10-min incubation, it also inhibited collagen-induced platelet aggregation with 50% inhibitory concentration (IC₅₀) of 70 nM. This is the first report of the active recombinant SVMP enzymes expressed in P. pastoris. The results suggest the significant roles of P-III SVMP in local and systemic pathology of envenomated patients. Inhibitors of this SVMP will be investigated in further studies to find a better treatment for viper bites.
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Pricing and availability is not currently available.