Merck
  • Therapeutic potential of amniotic-fluid-derived stem cells on liver fibrosis model in mice.
Therapeutic potential of amniotic-fluid-derived stem cells on liver fibrosis model in mice.
Taiwanese journal of obstetrics & gynecology (2014-07-16)
Shao-Yu Peng, Chih-Jen Chou, Po-Jen Cheng, I-Chen Ko, Yi-Jung Kao, Yu-Hsu Chen, Winston Teng-Kui Cheng, S W Steven Shaw, Shinn-Chih Wu
ABSTRACT
Liver fibrosis results from the wound healing response to chronic liver damage. Advanced liver fibrosis results in cirrhosis and liver failure, and liver transplantation is often the only option for effective therapy; however, the shortage of available donor livers limits this treatment. Thus, new therapies for advanced liver fibrosis are essential. Amniotic fluid contains an abundance of stem cells, which are derived from all three germ layers of the developing fetus. These cells do not induce teratomas in vivo and do not pose any ethical concerns. To generate liver fibrosis models, male ICR mice were treated with CCl4 via oral gavage for 4 weeks, and the serum levels of glutamate oxaloacetate transaminase, glutamate pyruvate transaminase, and albumin were higher than in the control group following chemical induction. To assess the potential of amniotic-fluid-derived stem cells (mAFSCs) to ameliorate liver fibrosis in vivo, mAFSCs were isolated from amniotic fluid of 13.5-day-old transgenic mice, which globally express the fluorescent protein, enhanced green fluorescent protein (EGFP), for tracing purposes (EGFP-mAFSCs). Single cells were injected via the mesentery (1 × 10(6) cells/mouse) of transplanted mice with liver fibrosis. Four weeks after EGFP-mAFSC transplantation, the serum glutamate oxaloacetate transaminase, glutamate pyruvate transaminase, and albumin levels of recipient mice in the EGFP-mAFSC-injected group were significantly decreased when compared with mice in the saline-injected group. Additionally, fibrotic tissues were evaluated using Masson's trichrome staining 4 weeks after cell transplantation. Shrinkage of the fibrotic area was observed in the EGFP-mAFSC-injected group. The tissue-repair effects were also confirmed by hydroxyproline content analysis. The possible repair mechanism from our data revealed that EGFP-mAFSCs may fuse with the recipient liver cells. Overall, EGFP-mAFSCs can ameliorate liver fibrosis in mice, thus providing insight into the future development of regenerative medicine.
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
Phosphomolybdic acid solution, spray reagent, 10% in ethanol
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
Phosphomolybdic acid solution, 20 wt. % in ethanol
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Phosphomolybdic acid solution, ready-to-use spray and plunge reagent, for chromatography
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
Indomethacin, Pharmaceutical Secondary Standard; Certified Reference Material
Indomethacin, European Pharmacopoeia (EP) Reference Standard
USP
Indomethacin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetic acid, LR, ≥99.5%
Sigma-Aldrich
Acetic acid, AR, ≥99.8%
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland