Merck
  • Inhibitory effects of a selective endothelin-A receptor antagonist YM598 on endothelin-1-induced potentiation of nociception in formalin-induced and prostate cancer-induced pain models in mice.
Inhibitory effects of a selective endothelin-A receptor antagonist YM598 on endothelin-1-induced potentiation of nociception in formalin-induced and prostate cancer-induced pain models in mice.
Journal of cardiovascular pharmacology (2005-04-20)
Hironori Yuyama, Akiko Koakutsu, Noriko Fujiyasu, Akira Fujimori, Shuichi Sato, Kumiko Shibasaki, Shohei Tanaka, Katsumi Sudoh, Masao Sasamata, Keiji Miyata
ABSTRACT
In some diseases in which endothelin-1 (ET-1) production increases (e.g. prostate cancer), ET-1 is considered to be involved in the generation of pain. In the present study, we investigated the effects of a selective endothelin-A receptor antagonist, YM598, on the nociception potentiated by ET-1 in formalin-induced and cancer inoculation-induced pain models in mice. The formalin-induced pain model was prepared by intraplantar injection of 0.7% formalin into the hind paws of ICR mice, and the cancer pain model was prepared by inoculation of the human prostate cancer cell line PPC-1 into the hind paws of severe combined immunodeficiency mice. Formalin caused a biphasic pain response and paw edema in the mouse hind paw. ET-1 (10 pmol/paw) potentiated these responses, and single oral administration of YM598 (0.3-3 mg/kg) significantly inhibited this ET-1-induced potentiation of nociception and paw edema. ET-1 (10 pmol/paw) also potentiated the pain response in prostate cancer-inoculated mice. Both YM598 and atrasentan (0.3-3 mg/kg) significantly inhibited the ET-1-induced potentiation of nociception. These results suggest that selective endothelin-A receptor antagonists relieve pain in patients with various diseases in which ET-1 production increases (e.g. prostate cancer).