- Isolation and culture of mouse primary pancreatic acinar cells.
Isolation and culture of mouse primary pancreatic acinar cells.
Journal of visualized experiments : JoVE (2013-08-28)
Johann Gout, Roxane M Pommier, David F Vincent, Bastien Kaniewski, Sylvie Martel, Ulrich Valcourt, Laurent Bartholin
PMID23979477
ABSTRACT
This protocol permits rapid isolation (in less than 1 hr) of murine pancreatic acini, making it possible to maintain them in culture for more than one week. More than 20 x 10(6) acinar cells can be obtained from a single murine pancreas. This protocol offers the possibility to independently process as many as 10 pancreases in parallel. Because it preserves acinar architecture, this model is well suited for studying the physiology of the exocrine pancreas in vitro in contrast to cell lines established from pancreatic tumors, which display many genetic alterations resulting in partial or total loss of their acinar differentiation.