- Role of substrates and products of PI 3-kinase in regulating activation of Rac-related guanosine triphosphatases by Vav.
Role of substrates and products of PI 3-kinase in regulating activation of Rac-related guanosine triphosphatases by Vav.
Mitogen stimulation of cytoskeletal changes and c-jun amino-terminal kinases is mediated by Rac small guanine nucleotide-binding proteins. Vav, a guanosine diphosphate (GDP)-guanosine triphosphate (GTP) exchange factor for Rac that stimulates the exchange of bound GDP for GTP, bound to and was directly controlled by substrates and products of phosphoinositide (PI) 3-kinase. The PI 3-kinase substrate phosphatidylinositol-4,5-bisphosphate inhibited activation of Vav by the tyrosine kinase Lck, whereas the product phosphatidylinositol-3,4,5-trisphosphate enhanced phosphorylation and activation of Vav by Lck. Control of Vav in response to mitogens by the products of PI 3-kinase suggests a mechanism for Ras-dependent activation of Rac.