Merck
A highly selective optical sensor for aniline recognition.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2009-10-17)
Liming Zhang, Bin Li
ABSTRACT
In this paper, an optical sensor, 2,6-bis(2-benzimidazolyl)pyridine, is reported to be highly selective towards aniline. A unique spectral response of 2,6-bis(2-benzimidazolyl)pyridine towards aniline is found, and a linear relationship between the sensor's emission intensity and the aniline's concentration is observed, making 2,6-bis(2-benzimidazolyl)pyridine a promising candidate for practicable optical sensors of aniline recognition.
Sigma-Aldrich
2,6-Bis(2-benzimidazolyl)pyridine
Pricing and availability is not currently available.