Merck
  • Isolation and chromosomal mapping of human glycogen synthase kinase-3 alpha and -3 beta encoding genes.
Isolation and chromosomal mapping of human glycogen synthase kinase-3 alpha and -3 beta encoding genes.
Genome (1998-11-11)
P C Shaw, A F Davies, K F Lau, M Garcia-Barcelo, M M Waye, S Lovestone, C C Miller, B H Anderton
ABSTRACT
Glycogen synthase kinase-3 (GSK-3) is a serine-threonine kinase that exists as two isoforms, alpha and beta, encoded by separate genes. Phosphorylation targets include a variety of cytoplasmic and nuclear proteins. Recent studies found that neurofilaments, amyloid precursor protein, and tau proteins are substrates of GSK-3 and that aberrant phosphorylation of these proteins is implicated in pathologies of the nervous system. To analyse the organisation of these two genes, a YAC library was screened by polymerase chain reaction, using primers specific for human GSK-3 alpha and GSK-3 beta cDNA. Two clones, 220 and 285 kb in size, containing the complete GSK-3 alpha coding sequence, and two clones, 365 and 285 kb in size, containing the 5' coding sequence of GSK-3 beta, were isolated. By somatic cell hybrid panel DNA amplification and radiation hybrid mapping, GSK-3 alpha was found to be located at 19q13.2. On the other hand, by somatic cell hybrid panel DNA amplification and fluorescence in situ hybridisation using the 285-kb YAC clone, GSK-3 beta was mapped to 3q13.3.