Merck
  • Efficient cross-coupling of functionalized arylzinc halides catalyzed by a nickel chloride-diethyl phosphite system.
Efficient cross-coupling of functionalized arylzinc halides catalyzed by a nickel chloride-diethyl phosphite system.
Organic letters (2005-10-21)
Andrei Gavryushin, Christiane Kofink, Georg Manolikakes, Paul Knochel
ABSTRACT
[reaction: see text] The combination of diethyl phosphite and DMAP as ligands for nickel in an 8:1 THF-N-ethylpyrrolidinone (NEP) mixture allows a very efficient cross-coupling reaction to be performed between various functionalized arylzinc halides and aryl bromides, triflates and activated chlorides. The reaction proceeds at 25 degrees C within 1-48 h and requires only 0.05 mol % of the nickel catalyst.
Sigma-Aldrich
Diethyl phosphite, technical grade, 94%