Merck
  • Synthesis and antibacterial activity of arylpiperazinyl oxazolidinones with diversification of the N-substituents.
Synthesis and antibacterial activity of arylpiperazinyl oxazolidinones with diversification of the N-substituents.
Bioorganic & medicinal chemistry letters (2004-07-01)
Sun-Young Jang, Young Hwan Ha, Seung Whan Ko, Wonku Lee, Jongkook Lee, Sunghoon Kim, Yong Woo Kim, Won Koo Lee, Hyun-Joon Ha
ABSTRACT
A series of 4-arylpiperazin-1-yl-3-phenyloxazolidin-2-one derivatives with diversification of the N-substituents such as methylene O-linked heterocycles, thioamide, dithiocarbamate, thiourea, and thiocarbamate were synthesized and evaluated as antibacterial agents. Their in vitro activities (MIC) were evaluated against MRSA and VRE resistant Gram-positive strains such as Staphylococcus and Enterococcus. Most of the compounds were more potent in vitro but less active in vivo than linezolid.
Sigma-Aldrich
1,1′-Thiocarbonyldi-2(1H)-pyridone, 97%
Pricing and availability is not currently available.