Merck
Visualization of arrestin recruitment by a G-protein-coupled receptor.
Nature (2014-07-22)
Arun K Shukla, Gerwin H Westfield, Kunhong Xiao, Rosana I Reis, Li-Yin Huang, Prachi Tripathi-Shukla, Jiang Qian, Sheng Li, Adi Blanc, Austin N Oleskie, Anne M Dosey, Min Su, Cui-Rong Liang, Ling-Ling Gu, Jin-Ming Shan, Xin Chen, Rachel Hanna, Minjung Choi, Xiao Jie Yao, Bjoern U Klink, Alem W Kahsai, Sachdev S Sidhu, Shohei Koide, Pawel A Penczek, Anthony A Kossiakoff, Virgil L Woods, Brian K Kobilka, Georgios Skiniotis, Robert J Lefkowitz
ABSTRACT
G-protein-coupled receptors (GPCRs) are critically regulated by β-arrestins, which not only desensitize G-protein signalling but also initiate a G-protein-independent wave of signalling. A recent surge of structural data on a number of GPCRs, including the β2 adrenergic receptor (β2AR)-G-protein complex, has provided novel insights into the structural basis of receptor activation. However, complementary information has been lacking on the recruitment of β-arrestins to activated GPCRs, primarily owing to challenges in obtaining stable receptor-β-arrestin complexes for structural studies. Here we devised a strategy for forming and purifying a functional human β2AR-β-arrestin-1 complex that allowed us to visualize its architecture by single-particle negative-stain electron microscopy and to characterize the interactions between β2AR and β-arrestin 1 using hydrogen-deuterium exchange mass spectrometry (HDX-MS) and chemical crosslinking. Electron microscopy two-dimensional averages and three-dimensional reconstructions reveal bimodal binding of β-arrestin 1 to the β2AR, involving two separate sets of interactions, one with the phosphorylated carboxy terminus of the receptor and the other with its seven-transmembrane core. Areas of reduced HDX together with identification of crosslinked residues suggest engagement of the finger loop of β-arrestin 1 with the seven-transmembrane core of the receptor. In contrast, focal areas of raised HDX levels indicate regions of increased dynamics in both the N and C domains of β-arrestin 1 when coupled to the β2AR. A molecular model of the β2AR-β-arrestin signalling complex was made by docking activated β-arrestin 1 and β2AR crystal structures into the electron microscopy map densities with constraints provided by HDX-MS and crosslinking, allowing us to obtain valuable insights into the overall architecture of a receptor-arrestin complex. The dynamic and structural information presented here provides a framework for better understanding the basis of GPCR regulation by arrestins.
Sigma-Aldrich
Adenosine 5′-Triphosphatase from porcine cerebral cortex, lyophilized powder, ≥0.3 units/mg protein, pH 7.8