Merck
  • Shell cross-linked micelles as cationic templates for the preparation of silica-coated nanoparticles: strategies for controlling the mean particle diameter.
Shell cross-linked micelles as cationic templates for the preparation of silica-coated nanoparticles: strategies for controlling the mean particle diameter.
Macromolecular rapid communications (2009-03-19)
Yuting Li, Jianzhong Du, Steven P Armes
ABSTRACT
The mean diameter of poly[2-(dimethylamino)ethyl methacrylate]-block-poly[2-(diisopropylamino)ethyl methacrylate] (PDMA-PDPA) diblock copolymer micelles can be easily adjusted from 27-155 nm (as measured by DLS) by either selective quaternisation of the PDMA block or by adding PDPA homopolymer prior to micellisation; these self-assembled nanostructures can be shell crosslinked with 1,2-bis-(2-iodoethoxy)ethane and subsequently used as templates for the preparation of silica-coated nanoparticles and, ultimately, hollow silica nanoparticles.
Sigma-Aldrich
1,2-Bis(2-iodoethoxy)ethane, 96%
Pricing and availability is not currently available.