Merck
  • Synthesis and cytotoxic activity of benzopyranoxanthone analogues of benz.
Synthesis and cytotoxic activity of benzopyranoxanthone analogues of benz.
Chemical & pharmaceutical bulletin (2001-06-20)
C Sittisombut, N Costes, S Michel, M Koch, F Tillequin, B Pfeiffer, P Renard, A Pierré, G Atassi
ABSTRACT
Condensation of 3-hydroxy-2-naphthalenecarboxylic acid with phloroglucinol afforded 1,3-dihydroxy-12H-benzo[b]xanthen-12-one. Construction of an additional dimethylpyran ring onto this skeleton, by alkylation with 3-chloro-3-methyl-1-butyne followed by Claisen rearrangement, gave access to a series of benzo[b]pyrano[2,3-i]xanthen-6-ones and benzo[b]pyrano[3,2-h]xanthen-7-ones related to psorospermine and benzo[b]acronycine. In contrast with what is observed in the pyridoacridone and benzopyridoacridone series, the linear benzo[b]-pyrano[2,3-i]xanthen-6-one derivatives were more potent than their angular benzo[b]pyrano[3,2-h]xanthen-7-one isomers. cis-3,4-Diacetoxy-5-methoxy-2,2-dimethyl-3,4-dihydro-2H,6H-benzo[b]pyrano[2,3-i]xanthen-6-one, the most active among the new compounds, was more potent than acronycine in inhibiting the proliferation of L1210 murine leukemia cells.
Sigma-Aldrich
3-Chloro-3-methyl-1-butyne, 97%
Pricing and availability is not currently available.