Merck
  • Characterization of volatile components and odor-active compounds in the oil of edible mushroom Boletopsis leucomelas.
Characterization of volatile components and odor-active compounds in the oil of edible mushroom Boletopsis leucomelas.
Journal of oleo science (2014-06-03)
Sota Nosaka, Mitsuo Miyazawa
ABSTRACT
The volatile oil from Boletopsis leucomelas (Pers.) Fayod was extracted by hydrodistillation with diethylether, and the volatile components of the oil were analyzed by gas chromatography-mass spectrometry. The oil contained 86 components, representing 87.5% of the total oil. The main components of the oil were linoleic acid (15.0%), phenylacetaldehyde (11.2%), and palmitic acid (9.4%). Furthermore, sulfur-containing compounds including 3-thiophenecarboxaldehyde, 2-acetylthiazole, S-methyl methanethiosulfonate, and benzothiazole were detected using gas chromatography-pulsed flame photometric detection. The odor components were evaluated by the odor activity value, and aroma extract dilution analysis was performed through gas chromatography-olfactometry analysis. The oil had a mushroom-like, fatty, and burnt odor. The main components contributing to the mushroom-like and fatty odor were hexanal, nonanal, 1-octen-3-ol, and (2E)-nonenal, while the burnt odor was due to furfuryl alcohol, benzaldehyde, 5-methyl furfural, 2,3,5-trimethylpyrazine, 2-acethylthiazole, and indole.
Sigma-Aldrich
2-Acetylthiazole, 99%