Merck
  • Levothyroxine treatment and pregnancy outcome in women with subclinical hypothyroidism undergoing assisted reproduction technologies: systematic review and meta-analysis of RCTs.
Levothyroxine treatment and pregnancy outcome in women with subclinical hypothyroidism undergoing assisted reproduction technologies: systematic review and meta-analysis of RCTs.
Human reproduction update (2013-01-19)
B Velkeniers, A Van Meerhaeghe, K Poppe, D Unuane, H Tournaye, P Haentjens
ABSTRACT
BACKGROUND Previous meta-analyses of observational data indicate that pregnant women with subclinical hypothyroidism have an increased risk of adverse pregnancy outcome. Potential benefits of levothyroxine (LT4) supplementation remain unclear, and no systematic review or meta-analysis of trial findings is available in a setting of assisted reproduction technologies (ART). METHODS Relevant trials published until August 2012 were identified by searching MEDLINE, EMBASE, Web of Knowledge, the Cochrane Controlled Trials Register databases and bibliographies of retrieved publications without language restrictions. RESULTS From 630 articles retrieved, we included three trials with data on 220 patients. One of these three trials stated 'live delivery' as outcome. LT4 treatment resulted in a significantly higher delivery rate, with a pooled relative risk (RR) of 2.76 (95% confidence limits 1.20-6.44; P = 0.018; I(2) = 70%), a pooled absolute risk difference (ARD) of 36.3% (3.5-69.0%: P = 0.030) and a summary number needed to treat (NNT) of 3 (1-28) in favour of LT4 supplementation. LT4 treatment significantly lowered miscarriage rate with a pooled RR of 0.45 (0.24-0.82; P = 0.010; I(2) = 26%), a pooled ARD of -31.3% (-48.2 to -14.5%: P < 0.001) and a summary NNT of 3 (2-7) in favour of LT4 supplementation. LT4 treatment had no effect on clinical pregnancy (RR 1.75; 0.90-3.38; P = 0.098; I(2) = 82%). In an ART setting, no data are available on the effects of LT4 supplementation on premature delivery, arterial hypertension, placental abruption or pre-eclampsia. CONCLUSIONS Our meta-analyses provide evidence that LT4 supplementation should be recommended to improve clinical pregnancy outcome in women with subclinical hypothyroidism and/or thyroid autoimmunity undergoing ART. Further research is needed to determine pregnancy outcome after close monitoring of thyroid function to maintain thyroid-stimulating hormone and free T4 levels within the trimester-specific reference ranges for pregnancy.
Product Description
Supelco
L-Thyroxine (T4) solution, 100 μg/mL in methanol with 0.1N NH3, ampule of 1 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.
USP
Levothyroxine, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
L-Thyroxine, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
L-Thyroxine, powder, BioReagent, suitable for cell culture
Pricing and availability is not currently available.