Merck
  • Aripiprazole-induced seizure in a 3-year-old child: a case report and literature review.
Aripiprazole-induced seizure in a 3-year-old child: a case report and literature review.
Clinical neuropharmacology (2013-01-22)
Farouq I Thabet, Rochelle T Sweis, Suja A Joseph
ABSTRACT
Aripiprazole is an atypical antipsychotic medication that is a partial dopamine D(2) and serotonin 5-hydroxytryptamine (1A) receptor agonist and 5-hydroxytryptamine (2A) receptor antagonist. It has a safer profile compared to other antipsychotic medications with regard to its effect on weight, glucose tolerance, prolactin level, and cardiac conduction. The common neurological adverse effects include headache, agitation, insomnia, sleepiness, and extrapyramidal symptoms. Seizures have not been reported in the pediatric population and only twice in adult patients. Here, we report a case of a healthy 3-year-old child who experienced prolonged lethargy, dystonia, and 2 witnessed seizures after incidental ingestion of 30 mg of aripiprazole. To our knowledge, this is the first reported case of aripiprazole-induced seizures in a child.
Product Description
Aripiprazole, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.