Merck
  • Occurrence, fate and effects of azoxystrobin in aquatic ecosystems: a review.
Occurrence, fate and effects of azoxystrobin in aquatic ecosystems: a review.
Environment international (2013-01-15)
Elsa Teresa Rodrigues, Isabel Lopes, Miguel Ângelo Pardal
ABSTRACT
The use of pesticides for crop protection may result in the presence of toxic residues in environmental matrices. In the aquatic environment, pesticides might freely dissolve in the water or bind to suspended matter and to the sediments, and might be transferred to the organisms' tissues during bioaccumulation processes, resulting in adverse consequences to non-target species. One such group of synthetic organic pesticides widely used worldwide to combat pathogenic fungi affecting plants is the strobilurin chemical group. Whereas they are designed to control fungal pathogens, their general modes of action are not specific to fungi. Consequently, they can be potentially toxic to a wide range of non-target organisms. The present work had the intent to conduct an extensive literature review to find relevant research on the occurrence, fate and effects of azoxystrobin, the first patent of the strobilurin compounds, in aquatic ecosystems in order to identify strengths and gaps in the scientific database. Analytical procedures and existing legislation and regulations were also assessed. Data gathered in the present review revealed that analytical reference standards for the most relevant environmental metabolites of azoxystrobin are needed. Validated confirmatory methods for complex matrices, like sediment and aquatic organisms' tissues, are very limited. Important knowledge of base-line values of azoxystrobin and its metabolites in natural tropical and estuarine/marine ecosystems is lacking. Moreover, some environmental concentrations of azoxystrobin found in the present review are above the Regulatory Acceptable Concentration (RAC) in what concerns risk to aquatic invertebrates and the No Observed Ecologically Adverse Effect Concentration (NOEAEC) reported for freshwater communities. The present review also showed that there are very few data on azoxystrobin toxicity to different aquatic organisms, especially in what concerns estuarine/marine organisms. Besides, toxicity studies mostly address azoxystrobin and usually neglect the more relevant environmental metabolites. Further work is also required in what concerns effects of exposure to multi-stressors, e.g. pesticide mixtures. Even though Log K(ow) for azoxystrobin and R234886, the main metabolite of azoxystrobin in water, are below 3, the bio-concentration factor and the bioaccumulation potential for azoxystrobin are absent in the literature. Moreover, no single study on bioaccumulation and biomagnification processes was found in the present review.
Product Description
Supelco
Azoxystrobin, PESTANAL®, analytical standard