Merck
  • Gramicidin S and polymyxins: the revival of cationic cyclic peptide antibiotics.
Gramicidin S and polymyxins: the revival of cationic cyclic peptide antibiotics.
Cellular and molecular life sciences : CMLS (2009-08-25)
Tatsushi Mogi, Kiyoshi Kita
ABSTRACT
Gramicidin S and polymyxins are small cationic cyclic peptides and act as potent antibiotics against Gram-negative and Gram-positive bacteria by perturbing integrity of the bacterial membranes. Screening of a natural antibiotics library with bacterial membrane vesicles identified gramicidin S as an inhibitor of cytochrome bd quinol oxidase and an alternative NADH dehydrogenase (NDH-2) and polymyxin B as an inhibitor of NDH-2 and malate: quinone oxidoreductase. Our studies showed that cationic cyclic peptide antibiotics have novel molecular targets in the membrane and interfere ligand binding on the hydrophobic surface of enzymes. Improvement of the toxicity and optimization of the structures and clinical uses are urgently needed for their effective application in combating drug-resistant bacteria.
Sigma-Aldrich
Gramicidin from Bacillus aneurinolyticus (Bacillus brevis), Linear polypeptide antibiotic complex. A mixture of gramicidins A, B, C, and D.
Pricing and availability is not currently available.
Gramicidin, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.