Merck
  • Effect of Crocus sativus L. (saffron) on coagulation and anticoagulation systems in healthy volunteers.
Effect of Crocus sativus L. (saffron) on coagulation and anticoagulation systems in healthy volunteers.
Phytotherapy research : PTR (2013-06-05)
Hossein Ayatollahi, Atefeh Ordoei Javan, Mohammad Khajedaluee, Masood Shahroodian, Hossein Hosseinzadeh
ABSTRACT
Saffron showed some effects on blood coagulation and platelet aggregation in in vitro and in vivo studies. In a clinical trial with a limited number volunteers, saffron tablets influenced on bleeding time. In this study, the effect of saffron on plasma level of fibrinogen, factor VII (as coagulant agent), C and S protein (as anti-coagulant agent), PT and PTT in a larger sample size was evaluated. The study was a double-blind, placebo-controlled study consisting of 1 week treatment with 200 mg and 400 mg saffron tablets. Sixty healthy volunteers (age range 20-50 years) were selected for the study. The volunteers were divided into three groups of 20 each. Group 1 received placebo; Groups 2 and 3 received 200 mg and 400 mg saffron tablets, respectively, for 7 days (1 tablet per day). Before and after 7 days treatment and also 1 month after that, blood samples were taken. The plasma levels of fibrinogen, factor VII, C and S protein, PT and PTT were evaluated. Statistical analysis showed no difference between groups for any of evaluated factors. This study rejected any effect of saffron with dose of 200 and 400 mg for 1 week on coagulant and anticoagulant system.
Product Description
Human coagulation factor VII concentrate BRP, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.