Merck
  • Potentially important nighttime heterogeneous chemistry: NO3 with aldehydes and N2O5 with alcohols.
Potentially important nighttime heterogeneous chemistry: NO3 with aldehydes and N2O5 with alcohols.
Physical chemistry chemical physics : PCCP (2011-04-22)
Richard Iannone, Song Xiao, Allan K Bertram
ABSTRACT
We report the first measurements of the reactive uptake of NO(3) with condensed-phase aldehydes. Specifically, we studied NO(3) uptake on solid tridecanal and the uptake on liquid binary mixtures containing tridecanal and saturated organic molecules (diethyl sebacate, dioctyl sebacate, and squalane) which we call matrix molecules. Uptake on the solid was shown to be efficient, where γ = (1.6 ± 0.8) × 10(-2). For liquid binary mixtures the reactivity of aldehyde depended on the matrix molecule. Assuming a bulk reaction, H(matrix)√(D(matrix)k(2°,aldehyde)) varied by a factor of 2.6, and assuming a surface reaction H(matrix)(S)K(matrix)(S)k(2°,aldehyde)(S) varied by a factor of 2.9, where H(matrix)√(D(matrix)k(2°,aldehyde)) and H(matrix)(S)K(matrix)(S)k(2°,aldehyde)(S) are constants extracted from the data using the resistor model. By assuming either a bulk or surface reaction, the atmospheric lifetimes for aldehydes were estimated to range from 1.9-7.5 h. We also carried out detailed studies of N(2)O(5) uptake kinetics on alcohols. We show that uptake coefficients of N(2)O(5) for five different organics at 293 K varied by more than 2 orders of magnitude, ranging from 3 × 10(-4) to 1.8 × 10(-2). We show that the uptake coefficients correlate with √(D(alcohol)(OH concentration)) but more work is needed with other alcohols to completely understand the dependence. Using this kinetic data we show that the atmospheric lifetime of alcohols with respect to N(2)O(5) heterogeneous chemistry can vary from 0.6-130 h, depending on the physical and chemical properties of the organic liquid.
Sigma-Aldrich
Tridecanal, technical grade, 90%
Pricing and availability is not currently available.