Merck
  • Isolation of elemental sulfur as a self-growth-inhibiting substance produced by Legionella pneumophila.
Isolation of elemental sulfur as a self-growth-inhibiting substance produced by Legionella pneumophila.
Applied and environmental microbiology (2002-09-27)
Hiroaki Inoue, Genji Kawano, Hiromichi Nagasawa, Shohei Sakuda
ABSTRACT
The addition of HP-20 resin to a medium could enhance the growth of Legionella species. Elemental sulfur was isolated as a self-growth-inhibiting substance produced endogenously by Legionella pneumophila from methanol extracts of the resins used to culture the bacterium. Elemental sulfur shows strong growth-inhibiting activity toward all Legionella species tested.
Product Description
Supelco
Diaion® HP-20, pkg of 1000 g
Supelco
Diaion® HP-20, pkg of 500 g