Merck
  • Accumulation of the mycotoxin patulin in the presence of gluconic acid contributes to pathogenicity of Penicillium expansum.
Accumulation of the mycotoxin patulin in the presence of gluconic acid contributes to pathogenicity of Penicillium expansum.
Molecular plant-microbe interactions : MPMI (2013-09-13)
Shiri Barad, Sigal Brown Horowitz, Ilana Kobiler, Amir Sherman, Dov Prusky
ABSTRACT
Penicillium expansum, the causal agent of blue mold rot, causes severe postharvest fruit maceration through secretion of D-gluconic acid (GLA) and secondary metabolites such as the mycotoxin patulin in colonized tissue. GLA involvement in pathogenicity has been suggested but the mechanism of patulin accumulation and its contribution to P. expansum pathogenicity remain unclear. The roles of GLA and patulin accumulation in P. expansum pathogenicity were studied using i) glucose oxidase GOX2-RNAi mutants exhibiting decreased GOX2 expression, GLA accumulation, and reduced pathogenicity; ii) IDH-RNAi mutants exhibiting downregulation of IDH (the last gene in patulin biosynthesis), reduced patulin accumulation, and no effect on GLA level; and iii) PACC-RNAi mutants exhibiting downregulation of both GOX2 and IDH that reduced GLA and patulin production. Present results indicate that conditions enhancing the decrease in GLA accumulation by GOX2-RNAi and PACC-RNAi mutants, and not low pH, affected patulin accumulation, suggesting GLA production as the driving force for further patulin accumulation. Thus, it is suggested that GLA accumulation may modulate patulin synthesis as a direct precursor under dynamic pH conditions modulating the activation of the transcription factor PACC and the consequent pathogenicity factors, which contribute to host-tissue colonization by P. expansum.
Sigma-Aldrich
Glucose Oxidase from Aspergillus niger, Type VII, lyophilized powder, ≥100,000 units/g solid (without added oxygen)
Sigma-Aldrich
Glucose Oxidase from Aspergillus niger, Type X-S, lyophilized powder, 100,000-250,000 units/g solid (without added oxygen)
Sigma-Aldrich
Glucose Oxidase from Aspergillus niger, Type II, ≥10,000 units/g solid (without added oxygen)