Merck
Synthesis of N-(purin-8-yl)arylamines.
Chemical research in toxicology (1993-07-01)
D H Swenson, K el-Bayoumy, G H Shuie, S S Hecht, E Fiala, F F Kadlubar, J P Freeman
ABSTRACT
Methods for direct synthesis of N-(purin-8-yl)arylamines were investigated. N-(Purin-8-yl)arylamines are adducts from reaction of electrophilic metabolites of arylamines with DNA and have not been readily available by direct synthesis. Ability to generate significant quantities of this class of DNA adduct by synthetic means would facilitate basic research in molecular toxicology. Two routes with common thiourea intermediates were developed for this purpose. In the first route, 6-hydroxy-2,4,5-triaminopyrimidine was caused to react in aqueous medium with dithiocarbamate derivatives of o-, m-, or p-toluidine to form corresponding 2,4-diamino-5-(tolylthioureido)aminopyrimidin-6-ones in 60-70% yields. The thiourea intermediates were converted to carbodiimides and isolated in 20-25% yields after treatment with HgO in dimethylformamide. The carbodiimides were cyclized at 125 degrees C in dimethylformamide under N2 for 45 h to yield N-(guanin-8-yl)(o-, m-, or p-)toluidines in 65-75% yields. In the second route, direct reaction of p-tolyl isothiocyanate with 6-hydroxy-2,4,5-triaminopyrimidine or with 4,5,6-triaminopyrimidine in dimethylformamide and triethylamine led to the formation of 2,4-diamino-5-(p-tolylthioureido)aminopyrimidin-6-one and 4,6-diamino-5-(tolylthioureido)aminopyrimidine, respectively, in 77-79% yields. Methylation of the two thiourea derivatives by methyl iodide in dimethylformamide gave the corresponding methylisothiuronium derivatives, which were isolated in 71% and 84% yields (as HI salts), respectively. Conversion of the methylisothiuronium derivatives to N-(guanin-8-yl)-p-toluidine or N-(adenin-8-yl)-p-toluidine was accomplished by heating in dimethylformamide for 5-7 h at 80-90 degrees C, in 70% and 62% yields, respectively.(ABSTRACT TRUNCATED AT 250 WORDS)
Sigma-Aldrich
p-Tolyl isothiocyanate, 97%