Merck
  • Oral DAV131, a charcoal-based adsorbent, inhibits intestinal colonization by β-lactam-resistant Klebsiella pneumoniae in cefotaxime-treated mice.
Oral DAV131, a charcoal-based adsorbent, inhibits intestinal colonization by β-lactam-resistant Klebsiella pneumoniae in cefotaxime-treated mice.
Antimicrobial agents and chemotherapy (2013-08-21)
Nathalie Grall, Laurent Massias, Thu Thuy Nguyen, Sakina Sayah-Jeanne, Nicolas Ducrot, Elisabeth Chachaty, Jean de Gunzburg, Antoine Andremont
ABSTRACT
Antibiotics excreted into the intestinal tract, such as broad-spectrum cephalosporins, disrupt the indigenous microflora, affect colonization resistance (CR), and promote intestinal colonization by resistant bacteria. We tested whether oral DAV131, a charcoal-based adsorbent, would prevent colonization by a cefotaxime (CTX)-resistant Klebsiella pneumoniae strain (PUG-2) in CTX-treated mice. Mice received CTX, saline, CTX and DAV131, or saline and DAV131 for 3 days before oral challenge with 10(6) CFU of PUG-2. The fecal CTX concentrations and counts of PUG-2 were assayed. Fecal CTX disappeared when DAV131 was given concomitantly with CTX (P < 0.05), and the area under the curve of PUG-2 fecal density was significantly reduced (P < 0.01). In conclusion, reducing intestinal antibiotic exposure with DAV131 may reduce colonization by resistant strains during treatment compared to treatment with CTX only. This might open new possibilities for decreasing the impact of antibiotics on the intestinal microbiota during treatments.
Product Description
Cefotaxime acid, European Pharmacopoeia (EP) Reference Standard