Merck
  • Quercetin prevents experimental glucocorticoid-induced osteoporosis: a comparative study with alendronate.
Quercetin prevents experimental glucocorticoid-induced osteoporosis: a comparative study with alendronate.
Canadian journal of physiology and pharmacology (2013-05-10)
Hoda Derakhshanian, Mahmoud Djalali, Abolghassem Djazayery, Keramat Nourijelyani, Sajad Ghadbeigi, Hamideh Pishva, Ahmad Saedisomeolia, Arash Bahremand, Ahmad Reza Dehpour
ABSTRACT
Glucocorticoid-induced osteoporosis (GIO) is the most common type of secondary osteoporosis. The aim of this study was to compare the efficacy of quercetin, a plant-derived flavonoid, with alendronate in the prevention of GIO. Fifty-six Sprague-Dawley rats were randomly distributed among 7 groups (8 rats per group) and treated for 6 weeks with one of the following: (i) normal saline; (ii) 40 mg methylprednisolone sodium succinate (MP)/kg body mass; (iii) MP + 40 μg alendronate/kg; (iv) MP + 50 mg quercetin/kg; (v) MP + 40 μg alendronate/kg + 50 mg quercetin/kg; (vi) MP + 150 mg quercetin/kg; and (vii) MP + 40 μg alendronate/kg + 150 mg quercetin/kg. MP and alendronate were injected subcutaneously and quercetin was administered by oral gavage 3 days a week. At the end of the study, femur breaking strength was significantly decreased as a consequence of MP injection. This decrease was completely compensated for in groups receiving 50 mg quercetin/kg plus alendronate, and 150 mg quercetin/kg with or without alendronate. Quercetin noticeably elevated osteocalcin as a bone formation marker, while alendronate did not show such an effect. In addition, administration of 150 mg quercetin/kg increased femoral trabecular and cortical thickness by 36% and 22%, respectively, compared with the MP-treated group. These data suggest that 150 mg quercetin/kg, alone or in combination with alendronate, can completely prevent GIO through its bone formation stimulatory effect.
Product Description
Methylprednisolone hydrogen succinate, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
USP
Methylprednisolone hemisuccinate, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.
Methylprednisolone hydrogen succinate for performance test, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.