Merck
  • Effects of pioglitazone versus glimepiride exposure on hepatocellular fat content in type 2 diabetes.
Effects of pioglitazone versus glimepiride exposure on hepatocellular fat content in type 2 diabetes.
Diabetes, obesity & metabolism (2013-04-12)
E Phielix, A Brehm, E Bernroider, M Krssak, C-H Anderwald, M Krebs, A I Schmid, P Nowotny, M Roden
ABSTRACT
Thiazoledinediones decrease blood glucose by their insulin-sensitizing properties. Here, we examined whether pioglitazone plus nateglinide (PIO) interferes with hepatocellular lipid (HCL) content and/or improves insulin sensitivity in well-controlled non-obese patients with type 2 diabetes mellitus (T2DM). Sixteen patients [body mass index (BMI): 28 ± 1 kg/m(2) ; HbA1c: 7.1 ± 0.6%] were studied in a randomized, double-blind, 12-week parallel group trial, whereas matched healthy humans [non-diabetic control subjects (CON), BMI: 26 ± 1 kg/m(2)] were studied once. Treatment with pioglitazone (30 mg/day) plus nateglinide (PIO arm) to control for glimepiride-induced insulin secretion was compared to treatment with glimepiride (2 mg/day) plus placebo (GLI arm). Multinuclei magnetic resonance spectroscopy (MRS) was combined with pancreatic normoglycaemic-two-step-insulin clamps and stable isotopes to assess glucose turnover, glucose transport/phosphorylation, HCL and intramyocellular lipid (IMCL) contents, non-esterified fatty acids (NEFA) and adipokines. At baseline, HCL was approximately 5.6-fold higher in T2DM (p < 0.05 vs. CON). This was paralleled by approximately doubled leptin : adiponectin ratios (p < 0.05). HCL decreased by approximately 39% (p < 0.05) after PIO and only tended to decrease after GLI (p = 0.12). Treatment with PIO did not affect leptin : adiponectin ratios, but slightly improved (p < 0.05) insulin-mediated NEFA suppression, which related to lower HCL. PIO further prevented the insulin-induced increase in IMCL content of soleus and tibialis anterior muscles. Peripheral and hepatic insulin sensitivity, glucose transport and glycaemic control did not change in both groups. Short-term, low-dose thiazolidendione treatment improves insulin sensitivity of lipolysis and HCL, without affecting muscle and liver insulin sensitivity. It appears that metabolic PIO action in T2DM is primarily mediated via a decline in HCL associated with greater sensitivity of lipolysis to insulin.
Product Description
N-[(cis-4-Isopropylcyclohexyl)carbonyl]-D-phenylalanine, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.