Merck
  • Curcumins promote monocytic gene expression related to β-amyloid and superoxide dismutase clearance.
Curcumins promote monocytic gene expression related to β-amyloid and superoxide dismutase clearance.
Neuro-degenerative diseases (2011-12-14)
J R Cashman, S Gagliardi, M Lanier, S Ghirmai, K J Abel, M Fiala
ABSTRACT
Neurodegenerative diseases are associated with accumulation of modified proteins or peptides including amyloid-β (Aβ) in Alzheimer's disease (AD), and misfolded superoxide dismutase-1 (SOD-1) in amyotrophic lateral sclerosis (ALS). Clearance of Aβ or SOD-1 by the innate immune system may be important for controlling or preventing disease onset. Curcumins restore Aβ phagocytosis by peripheral blood mononuclear cells (PBMCs) from AD patients and Aβ clearance with upregulation of key genes including MGAT3, vitamin D receptor (VDR) and Toll-like receptors (TLRs). Certain curcumins inhibit inflammatory processes of PBMCs from ALS patients. We developed an in vitro system using human monocytes from patients and monocytic cell lines (i.e. U-937, THP-1) for evaluating curcuminoid potency of innate immune cell stimulation. Bisdemethoxycurcumin and certain analogs potentiated MGAT3,VDR and TLR gene expression 3- to 300-fold in U-937 cells. The effect of curcumins on inflammation in monocytes from patients with ALS was examined. Recursive medicinal chemistry was applied to identify compounds that stimulate the innate immune system for use in the clearance of Aβ in AD and the reversal of neuroinflammation and defective SOD-1 accumulation in ALS.
Product Description
USP
Bisdesmethoxycurcumin, United States Pharmacopeia (USP) Reference Standard