Merck
  • Effects of dopexamine on the intestinal microvascular blood flow and leukocyte activation in a sepsis model in rats.
Effects of dopexamine on the intestinal microvascular blood flow and leukocyte activation in a sepsis model in rats.
Critical care (London, England) (2006-08-09)
Jurgen Birnbaum, Edda Klotz, Claudia D Spies, Bjorn Lorenz, Patrick Stuebs, Ortrud Vargas Hein, Matthias Grundling, Dragan Pavlovic, Taras Usichenko, Michael Wendt, Wolfgang J Kox, Christian Lehmann
ABSTRACT
The administration of dopexamine may constitute a therapeutical option to improve hepatosplanchnic perfusion in sepsis. In order to verify this hypothesis, we administered dopexamine in an experimental sepsis model in rats. This prospective, randomized, controlled laboratory study was conducted in 42 Wistar rats. The animals were divided into 3 groups. Group 1 (CON group) served as control group. The Animals of groups 2 (LPS Group) and 3 received an endotoxin infusion (20 mg/kgfor 15 min). In addition, in group 3 (DPX group) dopexamine was administered 0.5 microg/kg/minover 4 hours. One half of the animals of each group underwent studies of intestinal microvascular blood flow (IMBF) using laser Doppler fluxmetry. In the other half an intravital microscopic evaluation of the leukocyte endothelium cell interaction in the intestinal microcirculation was performed. Functional capillary denstity (FCD) in the intestinal mucosaand the circular as well as the longitudinal muscle layer was estimated. One hour after endotoxin challenge IMBF decreased significantly in the untreated LPS group to 51% compared to baseline (p<0.05). In DPX treated endotoxin animals we found significantly higher values at the level of CON group. The after endotoxin challenge impaired FCD was improved by dopexamine in the longitudinal (DPX + 33% vs. LPS; p <0.05) and in the circular muscle layer (DPX + 48% vs. LPS; p < 0.05) as a result of dopexamine administration. The administration of dopexamine reduced the count of firmly adherent leukocytes when compared to the untreated LPS group (-31%, p<0.05). TNF-alpha plasma levels were reduced by dopexamine infusion (LPS group 3637 +/- 553 pg/mL; DPXgroup 1933 +/- 201 pg/mL) one hour after endotoxin challenge. The administration of dopexamine improved IMBF and FCD as parameters of intestinal microcirculation and reduced leukocyte activation as a parameter of inflammation in experimental sepsis.
Product Description
Dopexamine dihydrochloride, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.